Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1444589_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444589_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444589_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
240038_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
240038_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700013726.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013726.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013726.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013731.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013731.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013731.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013730.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013730.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013730.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013732.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013732.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013732.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013728.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013728.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013728.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013729.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013729.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013729.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013727.mm.1 |
NM_001205095.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013727.mm.1 |
NM_001205095.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013727.mm.1 |
TC1700001571.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1444589_at |
- |
- |
Mouse Expression Set 430 |
1444589_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444589_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444589_PM_at |
- |
- |