Assay Details
| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163434.1 | NP_001156906.1 | 1 - 2 | 52 | 84 | |
| GenBank mRNA | AK076079.1 | - | 1 - 2 | 48 | 84 | |
| AK077463.1 | - | 1 - 2 | 52 | 84 | ||
| AK144650.1 | - | 1 - 2 | 48 | 84 | ||
| AK144742.1 | - | 1 - 2 | 48 | 84 | ||
| AK146647.1 | - | 1 - 2 | 51 | 84 | ||
| AK148539.1 | - | 1 - 2 | 51 | 84 | ||
| AK149447.1 | - | 1 - 2 | 51 | 84 | ||
| AK151647.1 | - | 1 - 2 | 50 | 84 | ||
| AK151806.1 | - | 1 - 2 | 51 | 84 | ||
| AK152020.1 | - | 1 - 2 | 48 | 84 | ||
| AK152958.1 | - | 1 - 2 | 51 | 84 | ||
| AK159719.1 | - | 1 - 2 | 48 | 84 | ||
| AK160821.1 | - | 52 | 84 | |||
| AK166207.1 | - | 1 - 2 | 48 | 84 | ||
| AK166739.1 | - | 1 - 2 | 48 | 84 | ||
| AK167011.1 | - | 1 - 2 | 48 | 84 | ||
| AK168012.1 | - | 1 - 2 | 52 | 84 | ||
| AK169034.1 | - | 1 - 2 | 52 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076079.1, AK146647.1, AK148539.1, AK149447.1, AK151647.1, AK151806.1, AK152020.1, AK152958.1, AK159719.1, AK166739.1, AK167011.1, AK168012.1, AK169034.1, NM_001163434.1 transcripts.
Set Membership: |
|

Hspa5
Genomic Map