Assay Details
| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009196.4 | NP_033222.1 | 1 - 2 | 166 | 98 | |
| GenBank mRNA | AF058055.1 | - | 1 - 2 | 147 | 98 | |
| AK033516.1 | - | 1 - 2 | 164 | 98 | ||
| AK079379.1 | - | 1 - 2 | 162 | 98 | ||
| AK088762.1 | - | 1 - 2 | 164 | 98 | ||
| AK161701.1 | - | 1 - 2 | 164 | 98 | ||
| AK162682.1 | - | 1 - 2 | 163 | 98 | ||
| AK166606.1 | - | 1 - 2 | 164 | 98 | ||
| BC014777.1 | - | 1 - 2 | 174 | 3499773 | 98 | |
| BY729217.1 | - | 1 - 2 | 170 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF058055.1, AK033516.1, AK079379.1, AK088762.1, AK161701.1, AK162682.1, AK166606.1, BC014777.1, NM_009196.4 transcripts.
Set Membership: |
|

Slc16a1
Genomic Map