Assay Details
| Gene Symbol: Nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145434.4 | NP_663409.2 | 1 - 2 | 664 | 92 | |
| GenBank mRNA | AF291821.1 | - | 1 - 2 | 488 | 92 | |
| AK041047.1 | - | 1 - 2 | 519 | 92 | ||
| AK135640.1 | - | 1 - 2 | 526 | 92 | ||
| AK137398.1 | - | 1 - 2 | 551 | 92 | ||
| AK137582.1 | - | 1 - 2 | 529 | 92 | ||
| AK146430.1 | - | 1 - 2 | 621 | 92 | ||
| AK154931.1 | - | 1 - 2 | 665 | 92 | ||
| AK155597.1 | - | 1 - 2 | 665 | 92 | ||
| BC008989.1 | - | 1 - 2 | 109 | 3585478 | 92 | |
| X86010.1 | - | 1 - 2 | 524 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr1d1
Genomic Map