Assay Details
| Gene Symbol: Rnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_005089_RNR1.0 | - | 547 | 94 | ||
More Information
Additional Information:
NC_005089 (strand:+; 70..1024); s-rRNA
Set Membership: |
|

Rnr1
Genomic Map