Assay Details
| Gene Symbol: A830082N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015526.2 | 1 | 2818 | 116 | ||
| GenBank mRNA | CD354304.1 | - | 1 | 401 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger with UFM1-specific peptidase domain | Zufsp | 2700019D07Rik | -35.9kb | Sense | Chr. 10 - 33919142 - 33951347 on NCBI Build GRCm38 |
| Downstream | RWD domain containing 1 | Rwdd1 |
0710001K08Rik, 2610002D06Rik, 2700069A07Rik, IH1 |
1.5kb | Sense | Chr. 10 - 33996555 - 34019616 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A830082N09Rik
Genomic Map