Assay Details
| Gene Symbol: Gm9199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027860.1 | 1 | 124 | 117 | ||
| GenBank mRNA | BE853472.1 | - | 1 | 127 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4606 | Gm4606 | -162.2kb | Anti-sense | Chr. 14 - 72828606 - 72863423 on NCBI Build GRCm38 | |
| Downstream | regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2 | Rcbtb2 |
2610028E02Rik, 2810420M18Rik, AI448345, AW240694, Chc1l, RC/BTB2 |
111.9kb | Sense | Chr. 14 - 73138773 - 73184055 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9199
Genomic Map