Assay Details
| Gene Symbol: 5033404E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033600.1 | 1 | 116 | 103 | ||
| GenBank mRNA | AK017151.1 | - | 328 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 30, member 10 | Slc30a10 | E130106K10Rik, Gm212 | -28.5kb | Sense | Chr. 1 - 185454848 - 185468762 on NCBI Build GRCm38 | 
| Downstream | lysophospholipase-like 1 | Lyplal1 | BC027340, Q96AVO | 589.7kb | Anti-sense | Chr. 1 - 186087732 - 186117310 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Detect Genomic DNA  Mus musculus  Both primers and probe map within a single exon | 
MicroArray Probes:
 Back To Top
 Back To Top
 5033404E19Rik
5033404E19Rik Genomic Map
Genomic Map Back To Top
 Back To Top