Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1420318_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200022496.mm.1 |
NR_038101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022496.mm.1 |
TC0200002753.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200022491.mm.1 |
NR_038101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022491.mm.1 |
TC0200002753.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1420318_PM_at |
- |
- |
Mouse Expression Set 430 |
1420318_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420318_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420318_at |
- |
- |