Assay Details
| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040264.1 | 1 - 2 | 137 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | striatin interacting protein 2 | Strip2 |
D330017J20Rik, Fam40b |
-25.6kb | Sense | Chr. 6 - 29916976 - 29959681 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700080G18 gene | 1700080G18Rik | 53.9kb | Anti-sense | Chr. 6 - 30047407 - 30049144 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Smkr-ps
Genomic Map