Assay Details
| Gene Symbol: Kantr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040269.2 | 1 - 2 | 131 | 106 | ||
| GenBank mRNA | AW213569.1 | - | 1 - 2 | 106 | 106 | |
| BB852892.1 | - | 1 - 2 | 109 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysine (K)-specific demethylase 5C | Kdm5c |
D930009K15Rik, Jarid1c, Smcx, mKIAA0234 |
-48.1kb | Anti-sense | Chr. X - 152233196 - 152279102 on NCBI Build GRCm38 |
| Downstream | TSPY-like 2 | Tspyl2 |
CDA1, CINAP, DENTT, DXBwg1396e, DXHXS1008E, E130307F10Rik |
9.4kb | Sense | Chr. X - 152336852 - 152342506 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Kantr
Genomic Map