Assay Details
| Gene Symbol: Gm2061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040305.1 | 1 - 2 | 535 | 82 | ||
| GenBank mRNA | AK138296.1 | - | 1 - 2 | 536 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 30, member 10 |
Slc30a10 |
E130106K10Rik, Gm212 |
Anti-sense | Chr. 1 - 185454848 - 185468762 on NCBI Build GRCm38 | |
| Downstream | NSA2 ribosome biogenesis homolog pseudogene | 5033404E19Rik | 41.7kb | Anti-sense | Chr. 1 - 185497306 - 185498014 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm2061
Genomic Map