Assay Details
| Gene Symbol: AI854517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040311.1 | 3 - 4 | 416 | 89 | ||
| GenBank mRNA | BM950091.1 | - | 3 - 4 | 322 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 9-3 | Mir9-3 |
Mirn9-3, mir-9-3, mmu-mir-9-3 |
-19.2kb | Sense | Chr. 7 - 79505264 - 79505353 on NCBI Build GRCm38 |
| Downstream | Rhesus blood group-associated C glycoprotein | Rhcg | BB065800 | 65.5kb | Anti-sense | Chr. 7 - 79593363 - 79617657 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AI854517
Genomic Map