| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
NR_040400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025008.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300025000.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025000.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025000.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
NR_040400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300025007.mm.1 |
TC0300003187.mm.1 |
MTA TC |