| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300024995.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024995.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024995.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024992.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024992.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024992.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024996.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024996.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024996.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024996.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024996.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024994.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024994.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024994.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024998.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024998.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024998.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024998.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024998.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024997.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024997.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024997.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024997.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024997.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024993.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024993.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024993.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024999.mm.1 |
NR_040398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024999.mm.1 |
NR_040398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024999.mm.1 |
TC0300003187.mm.1 |
MTA TC |