Assay Details
| Gene Symbol: Gm15401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040422.1 | 3 - 4 | 215 | 80 | ||
| GenBank mRNA | BY086664.1 | - | 3 - 4 | 215 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trans-golgi network protein | Tgoln1 |
AU015504, D6Ertd384e, TGN38, TGN38A, Ttgn1 |
-13.1kb | Anti-sense | Chr. 6 - 72608420 - 72617000 on NCBI Build GRCm38 |
| Downstream | potassium channel modulatory factor 1 | Kcmf1 |
1700094M07Rik, Debt91, Pmcf |
204.6kb | Anti-sense | Chr. 6 - 72841114 - 72899979 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15401
Genomic Map