Assay Details
| Gene Symbol: Gm15787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040430.1 | 1 - 2 | 191 | 89 | ||
| GenBank mRNA | CJ068547.1 | - | 1 - 2 | 154 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | checkpoint with forkhead and ring finger domains | Chfr |
5730484M20Rik, C230082M18, RNF116 |
-1.3kb | Anti-sense | Chr. 5 - 110135759 - 110171973 on NCBI Build GRCm38 |
| Downstream |
golgi autoantigen, golgin subfamily a, 3 |
Golga3 |
5330413L04, 5430416E01Rik, AI449376, AW490576, G1-499-14, Mea-2, Mea2, repro27 |
155.0 | Anti-sense | Chr. 5 - 110176659 - 110226470 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15787
Genomic Map