Assay Details
Gene Symbol: 1700112H15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040472.1 | 1 - 2 | 77 | 74 | ||
GenBank mRNA | AK007177.1 | - | 1 - 2 | 85 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY707265.1 | - | 76 | Chr. - - [-] on Build | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dual specificity phosphatase 10 | Dusp10 |
2610306G15Rik, AI158871, MKP-5, MKP5 |
-478.7kb | Anti-sense | Chr. 1 - 184034461 - 184075636 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930488B22 gene | 4930488B22Rik | 154.8kb | Anti-sense | Chr. 1 - 184712497 - 184718989 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
