Assay Details
| Gene Symbol: Apela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297554.1 | NP_001284483.1 | 2 - 3 | 299 | 122 | |
| GenBank mRNA | CA537316.1 | - | 1 - 2 | 191 | 122 | |
| CA540782.1 | - | 1 - 2 | 191 | 122 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif-containing 61 | Trim61 |
2czf61, AU018704, C85201, E330039K03Rik, Rnf35 |
-17.0kb | Sense | Chr. 8 - 65012976 - 65018688 on NCBI Build GRCm38 |
| Downstream | cDNA sequence BC030870 | BC030870 | 48.5kb | Sense | Chr. 8 - 65085613 - 65129537 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Apela
Genomic Map