Assay Details
| Gene Symbol: Gm10637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040697.1 | 1 - 2 | 194 | 71 | ||
| GenBank mRNA | AK139810.1 | - | 1 - 2 | 194 | 71 | |
| DV649137.1 | - | 1 - 2 | 183 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5118 | Gm5118 |
A330085E11, EG330819 |
-96.5kb | Anti-sense | Chr. 8 - 87093310 - 87101571 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930535O05 gene | 4930535O05Rik | 258.6kb | Anti-sense | Chr. 8 - 87458404 - 87459437 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139810.1 transcripts.
Set Membership: |
|

Gm10637
Genomic Map