Assay Details
| Gene Symbol: Gm10637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040697.1 | 4 - 5 | 575 | 62 | ||
| GenBank mRNA | DV649137.1 | - | 4 - 5 | 564 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5118 | Gm5118 |
A330085E11, EG330819 |
-78.8kb | Anti-sense | Chr. 8 - 87093310 - 87101571 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930535O05 gene | 4930535O05Rik | 277.6kb | Anti-sense | Chr. 8 - 87458404 - 87459437 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10637
Genomic Map