Assay Details
| Gene Symbol: Gm10637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040697.1 | 6 - 7 | 730 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5118 | Gm5118 |
A330085E11, EG330819 |
-68.2kb | Anti-sense | Chr. 8 - 87093310 - 87101571 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930535O05 gene | 4930535O05Rik | 282.6kb | Anti-sense | Chr. 8 - 87458404 - 87459437 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179983 | AK139810.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323589 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179983 | AK139810.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06584611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07147703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06832802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07188865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07328932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06701119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07406414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07160964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07147696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06706016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07160668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06832798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07160967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26033479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0800020257.mm.1 | NR_040697.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800020257.mm.1 | NR_040697.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800020257.mm.1 | TC0800002641.mm.1 | MTA TC |

Gm10637
Genomic Map