Assay Details
| Gene Symbol: Gm10745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040751.1 | 2 - 3 | 251 | 95 | ||
| GenBank mRNA | AK145331.1 | - | 2 - 3 | 252 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 16 | Snx16 |
4930522N22Rik, AI117577, AI747675 |
-1.4mb | Sense | Chr. 3 - 10417817 - 10440131 on NCBI Build GRCm38 |
| Downstream | predicted gene 2464 | Gm2464 | 1.6mb | Sense | Chr. 3 - 13470439 - 13471829 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0200000763.rn
Additional Information:
This assay is in the 5'UTR region of the AK145331.1 transcripts.
Set Membership: |
|

Gm10745
Genomic Map