Assay Details
| Gene Symbol: 4930546K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040754.1 | 2 - 3 | 195 | 89 | ||
| GenBank mRNA | AK133167.1 | - | 2 - 3 | 196 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39321 |
Gm39321 | -56.2kb | Anti-sense | Chr. 9 - 42142092 - 42145394 on NCBI Build GRCm38 | |
| Downstream |
sterol-C5-desaturase (fungal ERG3, delta-5-desaturase) homolog (S. cerevisae) |
Sc5d |
A830037K02, A830073K23Rik, Sc5dl |
45.9kb | Sense | Chr. 9 - 42251592 - 42264300 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930546K05Rik
Genomic Map