Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900009835.mm.1 |
NR_040763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009835.mm.1 |
NR_040763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009835.mm.1 |
TC0900001257.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009836.mm.1 |
NR_040763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009836.mm.1 |
NR_040763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009836.mm.1 |
TC0900001257.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009837.mm.1 |
NR_040763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009837.mm.1 |
NR_040763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009837.mm.1 |
TC0900001257.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009833.mm.1 |
NR_040763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009833.mm.1 |
NR_040763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009833.mm.1 |
TC0900001257.mm.1 |
MTA TC |