Assay Details
| Gene Symbol: 3300005D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045080.1 | 1 - 2 | 502 | 63 | ||
| GenBank mRNA | AK019455.1 | - | 1 - 2 | 341 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, DHHC domain containing 14 |
Zdhhc14 |
B530001K09Rik, New1cp |
-45.6kb | Sense | Chr. 17 - 5491946 - 5753891 on NCBI Build GRCm38 |
| Downstream | sorting nexin 9 | Snx9 |
2700073N08Rik, SDP1, SH3PX1 |
38.1kb | Sense | Chr. 17 - 5841380 - 5930711 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

3300005D01Rik
Genomic Map