Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700010690.mm.1 |
TC0700001210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010690.mm.1 |
NR_045098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010690.mm.1 |
NR_045098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010688.mm.1 |
TC0700001210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010688.mm.1 |
NR_045098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010688.mm.1 |
NR_045098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010687.mm.1 |
TC0700001210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010687.mm.1 |
NR_045098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010687.mm.1 |
NR_045098.1 |
Refseq |