Assay Details
| Gene Symbol: 4933427I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045160.1 | 3 - 4 | 367 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 7 | Pax7 | Pax-7 | -93.7kb | Sense | Chr. 4 - 139737059 - 139833026 on NCBI Build GRCm38 |
| Downstream | kelch domain containing 7A | Klhdc7a | B230308G19Rik | 35.0kb | Sense | Chr. 4 - 139962173 - 139968026 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933427I22Rik
Genomic Map