Assay Details
| Gene Symbol: 2610316D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045172.1 | 2 - 3 | 164 | 97 | ||
| GenBank mRNA | AK012035.1 | - | 2 - 3 | 164 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 3, ERATO Doi 751, expressed |
D3Ertd751e |
2810009O15Rik, 4930415G15Rik |
-3.6mb | Anti-sense | Chr. 3 - 41742606 - 41803292 on NCBI Build GRCm38 |
| Downstream | protocadherin 10 | Pcdh10 |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
12.1kb | Anti-sense | Chr. 3 - 45378398 - 45434579 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2610316D01Rik
Genomic Map