Assay Details
| Gene Symbol: 4931431B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045183.1 | 1 - 2 | 193 | 82 | ||
| GenBank mRNA | AK007137.1 | - | 1 - 2 | 195 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39091 |
Gm39091 | -9.2kb | Sense | Chr. 7 - 128181799 - 128184096 on NCBI Build GRCm38 | |
| Downstream | cytochrome c oxidase subunit VIa polypeptide 2 | Cox6a2 |
CoxVIaH, VIaH |
2.1kb | Sense | Chr. 7 - 128205435 - 128206366 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4931431B13Rik
Genomic Map