Assay Details
| Gene Symbol: Gm16336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045267.1 | 2 - 3 | 226 | 96 | ||
| GenBank mRNA | BE633821.1 | - | 2 - 3 | 226 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MRV integration site 1 | Mrvi1 |
BB115629, Irag, Ris1 |
Anti-sense | Chr. 7 - 110868257 - 110982908 on NCBI Build GRCm38 | |
| Downstream |
Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae) |
Ctr9 |
AA409336, Sh2bp1, Tsbp, Tsp, mKIAA0155 |
123.1kb | Sense | Chr. 7 - 111028951 - 111056377 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16336
Genomic Map