Assay Details
| Gene Symbol: 4732491K20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045290.1 | 1 - 2 | 316 | 96 | ||
| GenBank mRNA | AK029100.1 | - | 1 - 2 | 285 | 96 | |
| CJ159212.1 | - | 1 - 2 | 317 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase 4 | Map3k4 |
D17Rp17, D17Rp17e, MAPKKK4, MEKK 4, MTK1, Mek4b, Mekk4, RP17, Rp17a, Tas, mKIAA0213 |
-47.0 | Anti-sense | Chr. 17 - 12227621 - 12318806 on NCBI Build GRCm38 |
| Downstream | plasminogen | Plg |
AI649309, Pg |
51.4kb | Sense | Chr. 17 - 12378609 - 12419384 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4732491K20Rik
Genomic Map