Assay Details
| Gene Symbol: Rptoros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045312.1 | 3 - 4 | 341 | 69 | ||
| NR_045313.1 | 2 - 3 | 226 | 69 | |||
| GenBank mRNA | AK040159.1 | - | 3 - 4 | 333 | 69 | |
| AK086908.1 | - | 2 - 3 | 227 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11762 | Gm11762 | OTTMUSG00000004029 | -251.8kb | Anti-sense | Chr. 11 - 119548630 - 119569046 on NCBI Build GRCm38 |
| Downstream | charged multivesicular body protein 6 | Chmp6 | 2400004G01Rik | 87.5kb | Anti-sense | Chr. 11 - 119913446 - 119919552 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rptoros
Genomic Map