Assay Details
| Gene Symbol: Gm13582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045335.1 | 1 - 2 | 93 | 76 | ||
| GenBank mRNA | AK138134.1 | - | 1 - 2 | 94 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA binding motif, single stranded interacting protein 1 |
Rbms1 |
2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1 |
-5.2kb | Sense | Chr. 2 - 60750193 - 60963450 on NCBI Build GRCm38 |
| Downstream | TRAF family member-associated Nf-kappa B activator | Tank |
C86182, E430026L09Rik, I-TRAF |
603.4kb | Anti-sense | Chr. 2 - 61578561 - 61654171 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138134.1 transcripts.
Set Membership: |
|

Gm13582
Genomic Map