Assay Details
| Gene Symbol: C130060C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045355.1 | 2 - 3 | 350 | 98 | ||
| GenBank mRNA | AK048429.1 | - | 2 - 3 | 337 | 98 | |
| AK163425.1 | - | 2 - 3 | 351 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centrosomal protein 78 | Cep78 |
5730599I05Rik, D030027P05 |
-1.5kb | Anti-sense | Chr. 19 - 15955768 - 15985016 on NCBI Build GRCm38 |
| Downstream |
guanine nucleotide binding protein, alpha q polypeptide |
Gnaq |
1110005L02Rik, 6230401I02Rik, AA408290, AW060788, Dsk1, Dsk10, Galphaq, Gq, GqI |
121.9kb | Sense | Chr. 19 - 16132831 - 16387453 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100003304.rn
Set Membership: |
|

C130060C02Rik
Genomic Map