Assay Details
| Gene Symbol: 2900057B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045365.1 | 1 - 2 | 253 | 109 | ||
| GenBank mRNA | AK013712.1 | - | 1 - 2 | 256 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 7C | Zbtb7c |
B230208J24Rik, Kr-pok, Zbtb36 |
Anti-sense | Chr. 18 - 75820178 - 76148564 on NCBI Build GRCm38 | |
| Downstream | microRNA 6358 | Mir6358 |
Gm27830, mmu-mir-6358 |
72.1kb | Anti-sense | Chr. 18 - 76170552 - 76170652 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2900057B20Rik
Genomic Map