Assay Details
| Gene Symbol: 1700034I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045380.1 | 1 | 1912 | 131 | ||
| GenBank mRNA | AK006596.1 | - | 1 | 1913 | 131 | |
| AK077003.1 | - | 1 | 1871 | 131 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 6430590A07 gene | 6430590A07Rik | EG619719 | -5.6kb | Sense | Chr. 3 - 40876646 - 40894553 on NCBI Build GRCm38 |
| Downstream |
La ribonucleoprotein domain family, member 1B |
Larp1b |
1700108L22Rik, 4933421B21Rik, AA690246, Larp2 |
48.1kb | Anti-sense | Chr. 3 - 40950631 - 40977797 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700034I23Rik
Genomic Map