Assay Details
| Gene Symbol: 2010010A06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045394.1 | 1 - 2 | 135 | 109 | ||
| GenBank mRNA | AK008172.1 | - | 1 - 2 | 136 | 109 | |
| AK008334.1 | - | 1 - 2 | 136 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dymeclin | Dym |
1810041M12Rik, 4933427L07Rik, C030019K18Rik |
-1.7kb | Anti-sense | Chr. 18 - 75018699 - 75286966 on NCBI Build GRCm38 |
| Downstream | predicted gene 20544 | Gm20544 | 61.3kb | Sense | Chr. 18 - 75358638 - 75367201 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2010010A06Rik
Genomic Map