Assay Details
| Gene Symbol: 1700121L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045453.1 | 2 - 3 | 164 | 72 | ||
| GenBank mRNA | AK007232.1 | - | 2 - 3 | 165 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, DHHC domain containing 15 |
Zdhhc15 | 6030457O13Rik | -162.0kb | Anti-sense | Chr. X - 104536969 - 104671090 on NCBI Build GRCm38 |
| Downstream |
melanoma antigen, family E, 2 |
Magee2 |
9630059J11Rik, BB131182, Mage-e2 |
12.5kb | Anti-sense | Chr. X - 104854952 - 104857267 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0X00002033.rn
Set Membership: |
|

1700121L16Rik
Genomic Map