Assay Details
| Gene Symbol: 1810020O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045482.1 | 1 - 2 | 293 | 75 | ||
| GenBank mRNA | AK131825.1 | - | 1 - 2 | 294 | 75 | |
| BY707616.1 | - | 1 - 2 | 294 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E230015B07 gene | E230015B07Rik | -1.2kb | Anti-sense | Chr. 6 - 87672370 - 87679920 on NCBI Build GRCm38 | |
| Downstream | EF hand and coiled-coil domain containing 1 | Efcc1 | Ccdc48 | 40.0kb | Anti-sense | Chr. 6 - 87730820 - 87755912 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1810020O05Rik
Genomic Map