Assay Details
| Gene Symbol: 2810404M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045497.1 | 4 - 5 | 433 | 104 | ||
| GenBank mRNA | AK012985.1 | - | 4 - 5 | 433 | 104 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY712057.1 | - | 433 | Chr. - - [-] on Build | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FSHD region gene 1 | Frg1 | -609.5kb | Anti-sense | Chr. 8 - 41397454 - 41417118 on NCBI Build GRCm38 | |
| Downstream |
zinc finger protein 353, pseudogene |
Zfp353-ps | Zfp353 | 36.5kb | Anti-sense | Chr. 8 - 42081329 - 42083312 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2810404M03Rik
Genomic Map