Assay Details
| Gene Symbol: Gm21944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002880.1 | 2 | 119 | 97 | ||
| GenBank mRNA | DQ012048.1 | - | 2 | 119 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 21119 |
Gm21119 | -231.9kb | Sense | Chr. 8 - 20614405 - 20657119 on NCBI Build GRCm38 | |
| Downstream | defensin beta 33 | Defb33 |
BD-33, EG654453 |
3.6kb | Sense | Chr. 8 - 20892667 - 20897723 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm21944
Genomic Map