Assay Details
| Gene Symbol: D330023K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040334.1 | 1 | 220 | 122 | ||
| GenBank mRNA | AK043516.1 | - | 1 | 222 | 122 | |
| AK084630.1 | - | 1 | 188 | 122 | ||
| AK085418.1 | - | 1 | 174 | 122 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | formin binding protein 1 | Fnbp1 |
1110057E06Rik, 2210010H06Rik, FBP1, Fbp17 |
-10.0kb | Sense | Chr. 2 - 31026206 - 31142027 on NCBI Build GRCm38 |
| Downstream | neuronal calcium sensor 1 | Ncs1 |
9430075O15Rik, A730032G13Rik, AI836659, Freq, Mfreq, NCS-1 |
93.5kb | Anti-sense | Chr. 2 - 31245828 - 31295989 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D330023K18Rik
Genomic Map