Assay Details
| Gene Symbol: C330046G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040658.1 | 4 | 1024 | 102 | ||
| GenBank mRNA | AK082870.1 | - | 4 | 1025 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytoskeleton-associated protein 4 | Ckap4 |
5630400A09Rik, CLIMP-63, P63 |
-13.5kb | Sense | Chr. 10 - 84526305 - 84533888 on NCBI Build GRCm38 |
| Downstream | t-complex 11 (mouse) like 2 | Tcp11l2 | E430026E19Rik | 28.8kb | Anti-sense | Chr. 10 - 84576947 - 84614355 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C330046G13Rik
Genomic Map