Assay Details
| Gene Symbol: Gm11413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045450.1 | 3 | 1023 | 71 | ||
| GenBank mRNA | AK086792.1 | - | 3 | 1023 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 39B | Ttc39b |
1810054D07Rik, 9130422G05Rik, AI450603, AV103077, AW215585 |
-54.0kb | Sense | Chr. 4 - 83220301 - 83324270 on NCBI Build GRCm38 |
| Downstream |
small nuclear RNA activating complex, polypeptide 3 |
Snapc3 |
1810020H02Rik, 4930558A07Rik, 5031401C21Rik, AI414457, E030018J20Rik |
38.2kb | Anti-sense | Chr. 4 - 83417701 - 83467658 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm11413
Genomic Map