Assay Details
| Gene Symbol: 1700003D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045477.1 | 3 | 824 | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 1, regulatory (inhibitor) subunit 1B |
Ppp1r1b |
AU040756, DARPP-32, Darpp32 |
Anti-sense | Chr. 11 - 98348738 - 98357795 on NCBI Build GRCm38 | |
| Downstream | START domain containing 3 | Stard3 |
Mln64, es64 |
6.4kb | Anti-sense | Chr. 11 - 98358367 - 98381112 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700003D09Rik
Genomic Map