Assay Details
| Gene Symbol: Celf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244891.1 | NP_001231820.1 | 4 - 5 | 567 | 72 | |
| NM_001244903.1 | NP_001231832.1 | 3 - 4 | 514 | 72 | ||
| NM_017368.3 | NP_059064.2 | 4 - 5 | 666 | 72 | ||
| NM_198683.2 | NP_941955.1 | 3 - 4 | 514 | 72 | ||
| GenBank mRNA | AF267535.1 | - | 2 - 3 | 270 | 72 | |
| AF314172.1 | - | 2 - 3 | 108 | 72 | ||
| AJ007987.1 | - | 2 - 3 | 259 | 72 | ||
| AK014492.1 | - | 3 - 4 | 427 | 72 | ||
| AK143698.1 | - | 4 - 5 | 468 | 72 | ||
| AK156725.1 | - | 3 - 4 | 427 | 72 | ||
| AK161213.1 | - | 3 - 4 | 474 | 72 | ||
| AK214229.1 | - | 2 - 3 | 237 | 72 | ||
| BC021393.1 | - | 3 - 4 | 379 | 72 | ||
| BU056861.1 | - | 3 - 4 | 514 | 72 | ||
| BU923429.1 | - | 3 - 4 | 369 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celf1
Genomic Map