Assay Details
| Gene Symbol: Celf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244891.1 | NP_001231820.1 | 5 - 6 | 616 | 98 | |
| NM_001244903.1 | NP_001231832.1 | 4 - 5 | 563 | 98 | ||
| NM_017368.3 | NP_059064.2 | 5 - 6 | 715 | 98 | ||
| NM_198683.2 | NP_941955.1 | 4 - 5 | 563 | 98 | ||
| GenBank mRNA | AF267535.1 | - | 3 - 4 | 319 | 98 | |
| AF314172.1 | - | 3 - 4 | 157 | 98 | ||
| AJ007987.1 | - | 3 - 4 | 308 | 98 | ||
| AK014492.1 | - | 4 - 5 | 476 | 98 | ||
| AK143698.1 | - | 5 - 6 | 517 | 98 | ||
| AK156725.1 | - | 4 - 5 | 476 | 98 | ||
| AK161213.1 | - | 4 - 5 | 523 | 98 | ||
| AK214229.1 | - | 3 - 4 | 286 | 98 | ||
| BC021393.1 | - | 4 - 5 | 428 | 98 | ||
| BU923429.1 | - | 4 - 5 | 418 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celf1
Genomic Map