Assay Details
| Gene Symbol: Celf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244891.1 | NP_001231820.1 | 7 - 8 | 834 | 62 | |
| NM_001244903.1 | NP_001231832.1 | 6 - 7 | 781 | 62 | ||
| NM_017368.3 | NP_059064.2 | 7 - 8 | 933 | 62 | ||
| NM_198683.2 | NP_941955.1 | 6 - 7 | 781 | 62 | ||
| GenBank mRNA | AB050499.1 | - | 3 - 4 | 242 | 62 | |
| AF267535.1 | - | 5 - 6 | 537 | 62 | ||
| AF314172.1 | - | 5 - 6 | 375 | 62 | ||
| AJ007987.1 | - | 5 - 6 | 526 | 62 | ||
| AK014492.1 | - | 6 - 7 | 694 | 62 | ||
| AK143698.1 | - | 7 - 8 | 735 | 62 | ||
| AK156725.1 | - | 6 - 7 | 694 | 62 | ||
| AK218377.1 | - | 2 - 3 | 106 | 62 | ||
| BC021393.1 | - | 6 - 7 | 646 | 62 | ||
| X61451.1 | - | 2 - 3 | 229 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celf1
Genomic Map