Assay Details
| Gene Symbol: Tor2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152800.3 | NP_690013.1 | 1 - 2 | 215 | 71 | |
| GenBank mRNA | AK162719.1 | - | 1 - 2 | 159 | 71 | |
| AK171247.1 | - | 1 - 2 | 167 | 71 | ||
| BC003466.1 | - | 1 - 2 | 200 | 2649388 | 71 | |
| BC024469.1 | - | 1 - 2 | 178 | 4976022 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tor2a
Genomic Map